accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 24974-73-0

IUPAC Name: (3-chlorophenyl)methanesulfonyl chloride
Synonyms: (3-chlorophenyl)methanesulfonyl chloride
Molecular Weight (g/mol): 225.08
Molecular Formula: C7H6Cl2O2S
InChi Key: LXTGNVLBPVVMSL-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(CS(Cl)(=O)=O)=C1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)