CAS RN 249762-74-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ PDGFR Tyrosine Kinase Inhibitor II
The PDGFR Tyrosine Kinase Inhibitor II, also referenced under CAS 249762-74-1, controls the biological activity of PDGFR Tyrosine Kinase. This small molecule/inhibitor is primarily used for Phosphorylation and Dephosphorylation applications.