accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2498-66-0

IUPAC Name: 7,12-dihydrotetraphene-7,12-dione
Synonyms: benz A anthracene-7,12-dione
Molecular Weight (g/mol): 258.28
Molecular Formula: C18H10O2
InChi Key: LHMRXAIRPKSGDE-UHFFFAOYSA-N
SMILES: O=C1C2=CC=CC=C2C(=O)C2=C1C=CC1=CC=CC=C21
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results