accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH2

CAS RN 2499-95-8

IUPAC Name: hexyl prop-2-enoate
Synonyms: hexyl acrylate
Molecular Weight (g/mol): 156.23
Molecular Formula: C9H16O2
InChi Key: LNMQRPPRQDGUDR-UHFFFAOYSA-N
SMILES: CCCCCCOC(=O)C=C
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)