accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 250-84-0

IUPAC Name: thieno[2,3-b]thiophene
Synonyms: thieno[2,3-b]thiophene
Molecular Weight (g/mol): 140.22
Molecular Formula: C6H4S2
InChi Key: YHBTXTFFTYXOFV-UHFFFAOYSA-N
SMILES: S1C=CC2=C1SC=C2
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results