CAS RN 2501-02-2
CAS RN 2501-02-2
IUPAC Name:
1-nitro-4-[(1E)-2-(4-nitrophenyl)ethenyl]benzene
Synonyms:
4,4'-dinitrostilbene
Molecular Weight (g/mol):
270.24
Molecular Formula:
C14H10N2O4
InChi Key:
CLVIIRIMEIEKOQ-OWOJBTEDSA-N
SMILES:
[O-][N+](=O)C1=CC=C(\C=C\C2=CC=C(C=C2)[N+]([O-])=O)C=C1
1
–
1
of
1
results
1
4,4'-Dinitrostilbene 98.0+%, TCI America™
CAS: 2501-02-2 Molecular Formula: C14H10N2O4 Molecular Weight (g/mol): 270.24 MDL Number: MFCD00017043 InChI Key: CLVIIRIMEIEKOQ-OWOJBTEDSA-N PubChem CID: 5377860 ChEBI: CHEBI:79798 IUPAC Name: 1-nitro-4-[(E)-2-(4-nitrophenyl)ethenyl]benzene SMILES: C1=CC(=CC=C1C=CC2=CC=C(C=C2)[N+](=O)[O-])[N+](=O)[O-]