accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNCH3O

CAS RN 25016-09-5

IUPAC Name: 1,3-dimethyl-1H-pyrazole-5-carbaldehyde
Synonyms: 2,5-dimethylpyrazole-3-carbaldehyde
Molecular Weight (g/mol): 124.14
Molecular Formula: C6H8N2O
InChi Key: JWYFGNVBKRJGTN-UHFFFAOYSA-N
SMILES: CN1N=C(C)C=C1C=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)