accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 250161-45-6

IUPAC Name: 4-(1,3-oxazol-5-yl)benzoic acid
Synonyms: 4-(1,3-oxazol-5-yl)benzoic acid
Molecular Weight (g/mol): 189.17
Molecular Formula: C10H7NO3
InChi Key: MCPBXFWYUHYSIZ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1)C1=CN=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results