accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOOOOOHOHHOHOHOHOOHHOOHOHH2OH2OH2O

CAS RN 250249-75-3

IUPAC Name: 2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-3-[(3,4,5-trihydroxy-6-{[(3,4,5-trihydroxy-6-methyloxan-2-yl)oxy]methyl}oxan-2-yl)oxy]-4H-chromen-4-one trihydrate
Synonyms: rutin trihydrate
Molecular Weight (g/mol): 664.57
Molecular Formula: C27H36O19
InChi Key: NLLBWFFSGHKUSY-UHFFFAOYNA-N
SMILES: O.O.O.CC1OC(OCC2OC(OC3=C(OC4=CC(O)=CC(O)=C4C3=O)C3=CC(O)=C(O)C=C3)C(O)C(O)C2O)C(O)C(O)C1O
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)