accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 25026-34-0

IUPAC Name: 2-(4-chlorophenyl)acetyl chloride
Synonyms: 2-(4-chlorophenyl)acetyl chloride
Molecular Weight (g/mol): 189.04
Molecular Formula: C8H6Cl2O
InChi Key: UMQUIRYNOVNYPA-UHFFFAOYSA-N
SMILES: ClC(=O)CC1=CC=C(Cl)C=C1
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)