accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 25026-64-6

OOFCl

CAS RN 25026-64-6

IUPAC Name: 3-chloro-5-fluorobenzoate
Synonyms: 3-chloro-5-fluorobenzoate
Molecular Weight (g/mol): 173.55
Molecular Formula: C7H3ClFO2
InChi Key: QFMRANWPGGSNHS-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC(F)=CC(Cl)=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results