accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH3OHO

CAS RN 2503-46-0

IUPAC Name: 1-(4-hydroxy-3-methoxyphenyl)propan-2-one
Synonyms: 1-(4-hydroxy-3-methoxyphenyl)propan-2-one
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: LFVCJQWZGDLHSD-UHFFFAOYSA-N
SMILES: COC1=CC(CC(C)=O)=CC=C1O
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results