accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClNCOClCl

CAS RN 2505-31-9

IUPAC Name: 1,3,5-trichloro-2-isocyanatobenzene
Synonyms: 1,3,5-trichloro-2-isocyanatobenzene
Molecular Weight (g/mol): 222.45
Molecular Formula: C7H2Cl3NO
InChi Key: VDYWXVDWKFAUKE-UHFFFAOYSA-N
SMILES: ClC1=CC(Cl)=C(N=C=O)C(Cl)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results