accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 25054-53-9

IUPAC Name: 2H-1,3-benzodioxole-5-carbonyl chloride
Synonyms: 2H-1,3-benzodioxole-5-carbonyl chloride
Molecular Weight (g/mol): 184.58
Molecular Formula: C8H5ClO3
InChi Key: ZRSGZIMDIHBXIN-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC=C2OCOC2=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Boiling Point 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)