CAS RN 2506-41-4
IUPAC Name:
2-(chloromethyl)naphthalene
Synonyms:
2-(chloroethyl)naphthalene
Molecular Weight (g/mol):
176.64
Molecular Formula:
C11H9Cl
InChi Key:
MPCHQYWZAVTABQ-UHFFFAOYSA-N
SMILES:
ClCC1=CC=C2C=CC=CC2=C1
1
–
1
of
1
results
1
Sigma Aldrich 2-(Chloromethyl)naphthalene
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.