accessibility menu, dialog, popup

UserName

Viewing profile as user:
GeCH2OCH2OCH2OCH2O

CAS RN 25063-27-8

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 361.06
Molecular Formula: C16H32GeO4
InChi Key: MPVCRVXMJOZDGL-UHFFFAOYSA-N
SMILES: [Ge].[CH2]CCC[O-].[CH2]CCC[O-].[CH2]CCC[O-].[CH2]CCC[O-]
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)