accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2508-29-4

IUPAC Name: 5-hydroxypentan-1-aminium
Synonyms: 5-hydroxypentan-1-aminium
Molecular Weight (g/mol): 104.17
Molecular Formula: C5H14NO
InChi Key: LQGKDMHENBFVRC-UHFFFAOYSA-O
SMILES: [NH3+]CCCCCO
Molecular Weight (g/mol) 
  • (5)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (5)
  • (5)
  • (4)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)