CAS RN 25090-33-9
IUPAC Name:
3,4-dihydro-2H-pyran-5-carbaldehyde
Synonyms:
5,6-dihydro-4H-pyran-3-carbaldehyde
Molecular Weight (g/mol):
112.13
Molecular Formula:
C6H8O2
InChi Key:
WWZVSCJTMPYTLY-UHFFFAOYSA-N
SMILES:
O=CC1=COCCC1
1
–
1
of
1
results
1
3,4-Dihydro-2H-pyran-5-carbaldehyde, Thermo Scientific™
CAS: 25090-33-9 Molecular Formula: C6H8O2 Molecular Weight (g/mol): 112.128 MDL Number: MFCD00085019 InChI Key: WWZVSCJTMPYTLY-UHFFFAOYSA-N PubChem CID: 2736151 IUPAC Name: 3,4-dihydro-2H-pyran-5-carbaldehyde SMILES: C1CC(=COC1)C=O