accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 25100-12-3

IUPAC Name: hydrogen N-cyclohexylhydroxylamine chloride
Synonyms: hydrogen N-cyclohexylhydroxylamine chloride
Molecular Weight (g/mol): 151.63
Molecular Formula: C6H14ClNO
InChi Key: SSVAHXZUFFSFER-UHFFFAOYSA-N
SMILES: [H+].[Cl-].ONC1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results