accessibility menu, dialog, popup

UserName

Viewing profile as user:
ON

CAS RN 25115-74-6

IUPAC Name: 4-oxo-1-phenylcyclohexane-1-carbonitrile
Synonyms: 4-oxo-1-phenylcyclohexane-1-carbonitrile
Molecular Weight (g/mol): 199.25
Molecular Formula: C13H13NO
InChi Key: GKXOABVSZWCJJK-UHFFFAOYSA-N
SMILES: O=C1CCC(CC1)(C#N)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)