CAS RN 25134-15-0
CAS RN 25134-15-0
IUPAC Name:
(cyclopenta-2,4-dien-1-yl)trimethylsilane
Synonyms:
cyclopenta-2,4-dien-1-yltrimethylsilane
Molecular Weight (g/mol):
138.29
Molecular Formula:
C8H14Si
InChi Key:
VMFHCJPMKUTMMQ-UHFFFAOYSA-N
SMILES:
C[Si](C)(C)C1C=CC=C1
1
–
1
of
1
results
1
Trimethylsilylcyclopentadiene (mixture of isomers) 97.0+%, TCI America™
CAS: 25134-15-0 Molecular Formula: C8H14Si Molecular Weight (g/mol): 138.285 InChI Key: VMFHCJPMKUTMMQ-UHFFFAOYSA-N Synonym: Cyclopentadienyltrimethylsilane PubChem CID: 91288 IUPAC Name: cyclopenta-2,4-dien-1-yl(trimethyl)silane SMILES: C[Si](C)(C)C1C=CC=C1