accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 25148-85-0

IUPAC Name: disodium phenylphosphonate
Synonyms: disodium phenylphosphonate
Molecular Weight (g/mol): 202.06
Molecular Formula: C6H5Na2O3P
InChi Key: JOQAMSDLZYQHMX-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]P([O-])(=O)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results