accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 25152-52-7

IUPAC Name: alumanylidynestibane
Synonyms: aluminium antimonide
Molecular Weight (g/mol): 148.74
Molecular Formula: AlSb
InChi Key: LVQULNGDVIKLPK-UHFFFAOYSA-N
SMILES: [Al]#[Sb]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results