accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CO(E)(E)

CAS RN 25152-84-5

IUPAC Name: (2E,4E)-deca-2,4-dienal
Synonyms: decadienal trans, trans-2,4-decadienal
Molecular Weight (g/mol): 152.24
Molecular Formula: C10H16O
InChi Key: JZQKTMZYLHNFPL-BLHCBFLLSA-N
SMILES: CCCCC\C=C\C=C\C=O
Boiling Point 
  • (1)
  • (1)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (3)
  • (3)
Percent Purity 
  • (1)
  • (2)
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (3)