accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2516-34-9

IUPAC Name: cyclobutanamine
Synonyms: cyclobutylamine
Molecular Weight (g/mol): 71.12
Molecular Formula: C4H9N
InChi Key: KZZKOVLJUKWSKX-UHFFFAOYSA-N
SMILES: NC1CCC1
Boiling Point 
  • (2)
  • (1)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (5)
  • (2)
  • (6)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)