accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrSN

CAS RN 2516-40-7

IUPAC Name: 2-bromo-1,3-benzothiazole
Synonyms: 2-bromo-1,3-benzothiazole
Molecular Weight (g/mol): 214.08
Molecular Formula: C7H4BrNS
InChi Key: DRLMMVPCYXFPEP-UHFFFAOYSA-N
SMILES: BrC1=NC2=CC=CC=C2S1
Percent Purity 
  • (1)
  • (1)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)