accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3N

CAS RN 2516-47-4

IUPAC Name: cyclopropylmethanaminium
Synonyms: cyclopropylmethanaminium
Molecular Weight (g/mol): 72.13
Molecular Formula: C4H10N
InChi Key: IGSKHXTUVXSOMB-UHFFFAOYSA-O
SMILES: [NH3+]CC1CC1
Boiling Point 
  • (3)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (3)
  • (3)
  • (2)
  • (3)
  • (5)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (5)