accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CH3CH3CNOOOOOCH3CH3NOCH3H3C

CAS RN 2516-92-9

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 510.72
Molecular Formula: C28H50N2O6
InChi Key: XVJWWMHPPKXKRU-UHFFFAOYSA-N
SMILES: CC1(C)CC(CC(C)(C)N1[O])OC(=O)CCCCCCCCC(=O)OC1CC(C)(C)N([O])C(C)(C)C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results