accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 25167-70-8

IUPAC Name: 2,4,4-trimethylpent-1-ene
Synonyms: 2,4,4-trimethyl-1-pentene
Molecular Weight (g/mol): 112.22
Molecular Formula: C8H16
InChi Key: FXNDIJDIPNCZQJ-UHFFFAOYSA-N
SMILES: CC(=C)CC(C)(C)C
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)