accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOHNCl

CAS RN 2519-61-1

IUPAC Name: 2-(4-chloro-1H-indol-3-yl)acetic acid
Synonyms: 4-chloroindole-3-acetic acid 4-cl-iaa 4-Cl-IAA
Molecular Weight (g/mol): 209.63
Molecular Formula: C10H8ClNO2
InChi Key: WNCFBCKZRJDRKZ-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CNC2=CC=CC(Cl)=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results