accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 25190-06-1

IUPAC Name: (2S)-2-butoxybutan-1-ol
Synonyms: polytetrahydrofuran
Molecular Weight (g/mol): 146.23
Molecular Formula: C8H18O2
InChi Key: BJZYYSAMLOBSDY-QMMMGPOBSA-N
SMILES: CCCCO[C@@H](CC)CO
Molecular Weight (g/mol) 
  • (2)
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)