CAS RN 25194-52-9
IUPAC Name:
6-bromopyridine-2-carboxamide
Synonyms:
6-bromopyridine-2-carboxamide
Molecular Weight (g/mol):
201.02
Molecular Formula:
C6H5BrN2O
InChi Key:
HWRUOHMDYQEWOD-UHFFFAOYSA-N
SMILES:
NC(=O)C1=NC(Br)=CC=C1
1
–
1
of
1
results
1
Promotions Available
6-Bromopyridine-2-carboxamide, 97%
6-Bromopyridine-2-carboxamide, 97%
CAS: 25194-52-9 Molecular Formula: C6H5BrN2O Molecular Weight (g/mol): 201.02 MDL Number: MFCD00234154 InChI Key: HWRUOHMDYQEWOD-UHFFFAOYSA-N PubChem CID: 21961408 IUPAC Name: 6-bromopyridine-2-carboxamide SMILES: NC(=O)C1=NC(Br)=CC=C1