CAS RN 25194-67-6
IUPAC Name:
6-phenoxypyridin-3-amine
Synonyms:
6-phenoxypyridin-3-amine
Molecular Weight (g/mol):
186.21
Molecular Formula:
C11H10N2O
InChi Key:
DETKIRMPBJPJRQ-UHFFFAOYSA-N
SMILES:
NC1=CC=C(OC2=CC=CC=C2)N=C1
1
–
1
of
1
results
1
6-Phenoxy-3-pyridinamine, ≥95%, Thermo Scientific™
CAS: 25194-67-6 Molecular Formula: C11H10N2O Molecular Weight (g/mol): 186.214 MDL Number: MFCD01692449 InChI Key: DETKIRMPBJPJRQ-UHFFFAOYSA-N Synonym: 6-phenoxy-3-pyridinamine,3-amino-6-phenoxypyridine,5-amino-2-phenoxypyridine,pyridine, 5-amino-2-phenoxy,6-phenoxypyridine-3-amine,3-pyridinamine,6-phenoxy,3-pyridinamine, 6-phenoxy,6-phenoxy-pyridin-3-ylamine,6-phenoxy-3-pyridylamine,2-phenoxy-5-aminopyridine PubChem CID: 212856 IUPAC Name: 6-phenoxypyridin-3-amine SMILES: C1=CC=C(C=C1)OC2=NC=C(C=C2)N