accessibility menu, dialog, popup

UserName

Viewing profile as user:
FOFCl

CAS RN 252004-45-8

IUPAC Name: 4-chloro-2,6-difluorobenzaldehyde
Synonyms: 4-chloro-2,6-difluorobenzaldehyde
Molecular Weight (g/mol): 176.55
Molecular Formula: C7H3ClF2O
InChi Key: GTTVSBCPMJQRSP-UHFFFAOYSA-N
SMILES: FC1=CC(Cl)=CC(F)=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results