accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 252049-10-8

IUPAC Name: (2S)-6-(dimethylamino)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid hydrochloride
Synonyms: (2S)-6-(dimethylamino)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}hexanoic acid hydrochloride
Molecular Weight (g/mol): 432.95
Molecular Formula: C23H29ClN2O4
InChi Key: SJFAFKDBWNFBCC-BOXHHOBZSA-N
SMILES: Cl.CN(C)CCCC[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)