CAS RN 2524-04-1
CAS RN 2524-04-1
IUPAC Name:
O,O-diethyl chlorophosphonothioate
Synonyms:
O,O-diethyl chlorophosphonothioate
Molecular Weight (g/mol):
188.61
Molecular Formula:
C4H10ClO2PS
InChi Key:
KMJJJTCKNZYTEY-UHFFFAOYSA-N
SMILES:
CCOP(Cl)(=S)OCC
1
–
1
of
1
results
1
Diethyl Chlorothiophosphate 97.0+%, TCI America™
CAS: 2524-04-1 Molecular Formula: C4H10ClO2PS Molecular Weight (g/mol): 188.61 MDL Number: MFCD00009088 InChI Key: KMJJJTCKNZYTEY-UHFFFAOYSA-N PubChem CID: 17305 IUPAC Name: chloro-diethoxy-sulfanylidene-$l^{5}-phosphane SMILES: CCOP(=S)(OCC)Cl