accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 25245-29-8

CH3OIOCH3OCH3

CAS RN 25245-29-8

IUPAC Name: 5-iodo-1,2,3-trimethoxybenzene
Synonyms: 5-iodo-1,2,3-trimethoxybenzene
Molecular Weight (g/mol): 294.09
Molecular Formula: C9H11IO3
InChi Key: IWPMQXOVTMABLF-UHFFFAOYSA-N
SMILES: COC1=CC(I)=CC(OC)=C1OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results