accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOIBr

CAS RN 25252-00-0

IUPAC Name: 2-bromo-5-iodobenzoic acid
Synonyms: 2-bromo-5-iodobenzoic acid
Molecular Weight (g/mol): 326.92
Molecular Formula: C7H4BrIO2
InChi Key: QPKKBDSNZFSSOD-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(I)=CC=C1Br
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results