accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOCH3FFFK

CAS RN 252726-24-2

IUPAC Name: potassium (4-acetylphenyl)trifluoroboranuide
Synonyms: potassium (4-acetylphenyl)trifluoroboranuide
Molecular Weight (g/mol): 226.05
Molecular Formula: C8H7BF3KO
InChi Key: JRUXIRSHKHCTTK-UHFFFAOYSA-N
SMILES: [K+].CC(=O)C1=CC=C(C=C1)[B-](F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results