accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 25276-70-4

IUPAC Name: pentadecane-1-thiol
Synonyms: pentadecane-1-thiol
Molecular Weight (g/mol): 244.48
Molecular Formula: C15H32S
InChi Key: IGMQODZGDORXEN-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCS

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results