accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2528-38-3

CH3OPOOCH3OCH3

CAS RN 2528-38-3

IUPAC Name: tripentyl phosphate
Synonyms: tripentyl phosphate
Molecular Weight (g/mol): 308.4
Molecular Formula: C15H33O4P
InChi Key: QJAVUVZBMMXBRO-UHFFFAOYSA-N
SMILES: CCCCCOP(=O)(OCCCCC)OCCCCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results