accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COPOOCH3OCH3

CAS RN 2528-39-4

IUPAC Name: trihexyl phosphate
Synonyms: trihexyl phosphate
Molecular Weight (g/mol): 350.48
Molecular Formula: C18H39O4P
InChi Key: SFENPMLASUEABX-UHFFFAOYSA-N
SMILES: CCCCCCOP(=O)(OCCCCCC)OCCCCCC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results