accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFFFFFFFFFFFH2C

CAS RN 25291-17-2

IUPAC Name: 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooct-1-ene
Synonyms: 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooct-1-ene
Molecular Weight (g/mol): 346.09
Molecular Formula: C8H3F13
InChi Key: FYQFWFHDPNXORA-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C=C
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)