accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 252916-29-3

IUPAC Name: 3-(2,4-dimethyl-5-{[(3E)-2-oxo-2,3-dihydro-1H-indol-3-ylidene]methyl}-1H-pyrrol-3-yl)propanoic acid
Synonyms: 3-(2,4-dimethyl-5-{[(3E)-2-oxo-1H-indol-3-ylidene]methyl}-1H-pyrrol-3-yl)propanoic acid
Molecular Weight (g/mol): 310.35
Molecular Formula: C18H18N2O3
InChi Key: NHFDRBXTEDBWCZ-NTEUORMPSA-N
SMILES: CC1=C(CCC(O)=O)C(C)=C(N1)\C=C1\C(=O)NC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results