CAS RN 2530-10-1
CAS RN 2530-10-1
IUPAC Name:
1-(2,5-dimethylthiophen-3-yl)ethan-1-one
Synonyms:
1-(2,5-dimethylthiophen-3-yl)ethanone
Molecular Weight (g/mol):
154.23
Molecular Formula:
C8H10OS
InChi Key:
PUSJAEJRDNPYKM-UHFFFAOYSA-N
SMILES:
CC(=O)C1=C(C)SC(C)=C1
1
–
1
of
1
results
1
Thermo Scientific Chemicals 3-Acetyl-2,5-dimethylthiophene, 99%
CAS: 2530-10-1 MDL Number: MFCD00009763