accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 25301-02-4

IUPAC Name: 4-(2,4,4-trimethylpentan-2-yl)phenol; formaldehyde; oxirane
Synonyms: tyloxapol
Molecular Weight (g/mol): 280.41
Molecular Formula: C17H28O3
InChi Key: MDYZKJNTKZIUSK-UHFFFAOYSA-N
SMILES: C=O.C1CO1.CC(C)(C)CC(C)(C)C1=CC=C(O)C=C1
Physical Form 
  • (2)
  • (10)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
Grade 
  • (5)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (10)
  • (10)
  • (2)
  • (5)
  • (2)
  • (10)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (5)
  • (5)