accessibility menu, dialog, popup

UserName

Viewing profile as user:
PdPPClClPhPhPhPh

CAS RN 253157-79-8

IUPAC Name: [2'-(diphenylphosphanyl)-[1,1'-binaphthalen]-2-yl]diphenylphosphane; dichloropalladiumbis(ylium)
Synonyms: binap; dichloropalladiumbis(ylium)
Molecular Weight (g/mol): 800.01
Molecular Formula: C44H32Cl2P2Pd
InChi Key: NOOYAPWTNFURJA-UHFFFAOYSA-L
SMILES: Cl[Pd++]Cl.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=C(C2=CC=CC=C2C=C1)C1=C(C=CC2=CC=CC=C12)P(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results