accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNOBr

CAS RN 253273-90-4

IUPAC Name: 3-[3-(bromomethyl)phenyl]-5-methyl-1,2,4-oxadiazole
Synonyms: 3-[3-(bromomethyl)phenyl]-5-methyl-1,2,4-oxadiazole
Molecular Weight (g/mol): 253.1
Molecular Formula: C10H9BrN2O
InChi Key: CERZNQPNTHWEAD-UHFFFAOYSA-N
SMILES: CC1=NC(=NO1)C1=CC(CBr)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results