accessibility menu, dialog, popup

UserName

Viewing profile as user:
Br(S)

CAS RN 2534-77-2

IUPAC Name: (2S)-2-bromobicyclo[2.2.1]heptane
Synonyms: (2S)-2-bromobicyclo[2.2.1]heptane
Molecular Weight (g/mol): 175.07
Molecular Formula: C7H11Br
InChi Key: QXYOAWHKJRWNID-AHXFUIDQSA-N
SMILES: Br[C@H]1CC2CCC1C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results