CAS RN 253429-19-5
IUPAC Name:
7-bromo-5-fluoro-1-benzofuran
Synonyms:
7-bromo-5-fluoro-1-benzofuran
Molecular Weight (g/mol):
215.02
Molecular Formula:
C8H4BrFO
InChi Key:
TYBMMMOEVBNTDU-UHFFFAOYSA-N
SMILES:
FC1=CC(Br)=C2OC=CC2=C1
1
–
1
of
1
results
1
Promotions Available
7-Bromo-5-fluorobenzo[b]furan, 97%
7-Bromo-5-fluorobenzo[b]furan, 97%
CAS: 253429-19-5 Molecular Formula: C8H4BrFO Molecular Weight (g/mol): 215.021 MDL Number: MFCD09056780 InChI Key: TYBMMMOEVBNTDU-UHFFFAOYSA-N PubChem CID: 22144816 IUPAC Name: 7-bromo-5-fluoro-1-benzofuran SMILES: C1=COC2=C(C=C(C=C21)F)Br